Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Protease Inhibitor Cocktail VII, His-Tag Sequences, 1 Vial
SDP

Catalog No. 50488962
Encompass_Preferred

Mixture of five protease inhibitors for purification of proteins containing hexa histidine tagged sequences. Supplied in 1ml DMSO. Will inhibit proteases extracted from 10g of cells.

Contains:

AEBSF
Bestatin
E-64
Pepstatin A
Phosphoromidon

Specificity:
Serine Proteases
Cysteine Proteases
Aspartic Proteases
Aminopeptidoses and Metalloproteases

Catalog No. 50-488-962 Supplier Research Products International Corp Supplier No. P514001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mixture of five protease inhibitors for purification of proteins containing hexa histidine tagged sequences. Supplied in 1ml DMSO. Will inhibit proteases extracted from 10g of cells.

Contains:

AEBSF
Bestatin
E-64
Pepstatin A
Phosphoromidon

Specificity:
Serine Proteases
Cysteine Proteases
Aspartic Proteases
Aminopeptidoses and Metalloproteases

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.