Learn More
Rockland Immunochemicals Protease S. aureus 1 mg
SDP
Catalog No. 501943978
Encompass_Preferred
Protease S. aureus V8 (Endoproteinase-Glu-C) specifically cleaves peptide bonds on the COOH-terminal side of either aspartic or glutamic acids. In the presence of ammonium the enzyme specificity is limited to glutamic sites. It has a molecular weight of 27 000 daltons and optimum pH's of 4.0 and 7.8 with hemoglobin as the substrate. Protease S. aureus V8 is inhibited by diisopropylfluorophosphate and monovalent anions such as F- Cl- CH3COO- and NO3. Enzyme activity is determined by the casein digestion assay. Protease S. aureus is ideal for investigators involved in enzyme and infectious diease research.
Catalog No. 50-194-3978 Supplier Rockland Immunochemicals Supplier No. MB1140001
Protease S. aureus V8 (Endoproteinase-Glu-C) specifically cleaves peptide bonds on the COOH-terminal side of either aspartic or glutamic acids. In the presence of ammonium the enzyme specificity is limited to glutamic sites. It has a molecular weight of 27 000 daltons and optimum pH's of 4.0 and 7.8 with hemoglobin as the substrate. Protease S. aureus V8 is inhibited by diisopropylfluorophosphate and monovalent anions such as F- Cl- CH3COO- and NO3. Enzyme activity is determined by the casein digestion assay. Protease S. aureus is ideal for investigators involved in enzyme and infectious diease research.