Learn More
Worthington Biochemical Corporation PROTEASE S AUREUS STRAIN V8
SDP
Catalog No. NC9007494
Encompass
Protease, S. Aureus (Endoproteinase Glu-C), 5mg, Code: STAP, Form: Lyophilized powder, Storage Temperature: 2degree to 8degreeC, Source: S. aureus V8, Minimum Activity: > = 500U/mg dry weight, Stability: 12 months
Catalog No. NC9007494 Supplier Worthington Biochemical Corporation Supplier No. LS003605
Protease, S. Aureus (Endoproteinase Glu-C), 5mg, Code: STAP, Form: Lyophilized powder, Storage Temperature: 2degree to 8degreeC, Source: S. aureus V8, Minimum Activity: > = 500U/mg dry weight, Stability: 12 months