Learn More
Biotium Protease Substrates Enzyme Substrates Protease Substrates Chemical Reagents 1/EA
Supplier:  Biotium 2909010000U
Encompass_Preferred
This kit contains recombinant TEV Protease and Control Substrate both expressed from E coli along with 10X Reaction Buffer All kit components are supplied in solution and can be used to cleave affinity tags from fusion proteins using the TEV protease recognition site Unit size 10000 U
Catalog No. 50-005-01185