Learn More
Biocare Medical, LLC Protease XXIV
SDP
Catalog No. 5083234
Encompass_Preferred
The staining pattern and intensity of signal for some IHC and ISH procedures can be improved by using an enzymatic pretreatment of the tissue sections. Digestive enzyme pretreatment is recommended prior to the addition of the primary antibody and/or probe. Always consult the protocol supplied with each primary antibody for optimal digestion conditions. It can also be used on an automated staining system.
Catalog No. 50-832-34 Supplier Biocare Medical, LLC Supplier No. PR960KG15
The staining pattern and intensity of signal for some IHC and ISH procedures can be improved by using an enzymatic pretreatment of the tissue sections. Digestive enzyme pretreatment is recommended prior to the addition of the primary antibody and/or probe. Always consult the protocol supplied with each primary antibody for optimal digestion conditions. It can also be used on an automated staining system.