Learn More
Enzo Life Sciences Proteasome purification kit (10 tests)
SDP
Catalog No. 502011565
Encompass_Preferred
This kit provides a gentle and rapid means to purify proteasome complexes from biological samples using an agarose immobilised 20S subunit specific proteasome antibody, without the need for genetic modification of the expressed proteasome. Captured proteasomes are eluted under denaturing conditions prior to analysis by Western blotting or characterisation by proteomic methods.
Catalog No. 50-201-1565 Supplier Enzo Life Sciences Supplier No. BMLPW1075A0001
This kit provides a gentle and rapid means to purify proteasome complexes from biological samples using an agarose immobilised 20S subunit specific proteasome antibody, without the need for genetic modification of the expressed proteasome. Captured proteasomes are eluted under denaturing conditions prior to analysis by Western blotting or characterisation by proteomic methods.