Learn More
Enzo Life Sciences Proteasome activator 11S gamma subunit polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81900100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 70-85 of human and mouse Ki autoantigen/PA28γ. Species reactivity: Human, Mouse. Specificity: Recognizes the γ subunit of proteasome activator 11S. Applications: IHC, WB. Formulation: Liquid. Antiserum containing 10mM sodium azide.
Catalog No. 50-201-1592