Learn More
Enzo Life Sciences 19S Proteasome (ADRM1) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2960100
Encompass_Preferred
Immunogen: Recombinant human ADRM1 (aa 2-407). Species reactivity: Human. Applications: ELISA, ICC, IHC (FS), IHC (PS), WB. Formulation: Liquid. In PBS, pH 7.4, containing 0.02% sodium azide and 50% glycerol.
Catalog No. 50-201-2836