Learn More
Enzo Life Sciences Proteasome 19S Rpn10 / S5a subunit (human), (recombinant) (GST-tag) (agarose immobilized) (0.5ML)
SDP
Supplier:  Enzo Life Sciences BMLUW86350500
Encompass_Preferred
Alternative name: 26S proteasome non-ATPase regulatory subunit 4, Antisecretory factor 1, Multiubiquitin chain-binding protein. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. The resin is supplied as a slurry in 50mM TRIS, pH 7.5, containing 150mM sodium chloride, 50% glycerol, 1mM DTT and 0.01% sodium azide. Source: GST-S5a (human Rpn10) is covalently coupled to agarose at a concentration of 0.5mg protein/ml of settled resin. Long Term Storage: +4°C.
Catalog No. 50-201-2289