Learn More
Enzo Life Sciences Proteasome 20S alpha7 subunit monoclonal antibody (MCP72) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81100025
Encompass_Preferred
Clone: MCP72. Host: Mouse. Isotype: IgG1. Immunogen: Dinitrophenylated human placenta derived proteasomes. UniProt: P25788. Source: Purified from hybridoma tissue culture supernatant. Species Reactivity: Human, Rat, Yeast, Rabbit, Arthropod. Specificity: Recognizes the α7 subunit of the 20S proteasome. Applications: IHC, WB. Purity: Partially purified ascites. Formulation: Liquid. In PBS containing 10mM sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. NC1781148