Learn More
Enzo Life Sciences Proteasome 20S alpha6 subunit monoclonal antibody (MCP20) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81000100
Encompass_Preferred
Clone: MCP20. Host: Mouse. Isotype: IgG1. Immunogen: Human placenta derived proteasomes. UniProt: P25786. Source: Purified from ascites. Species Reactivity: Human, Rabbit. Specificity: Recognizes the α6 subunit of proteasome 20S. Crossreactivity: Does not cross-react with rat or mouse protein. Applications: IHC, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 10mM sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. NC0217734