Learn More
Enzo Life Sciences Proteasome 20S assay kit (1x96wells)
SDP
Catalog No. 502010300
Encompass_Preferred
A QUANTIZYME Assay System. Fluorogenic, non-radioactive assay designed to measure chymotrypsin-like protease activity of purified 20S proteasome. This 96-well assay is useful for screening inhibitors and modulators of the 20S proteasome. The kit includes purified, human erythrocyte 20S proteasome (20µg). The Suc-LLVY-AMC fluorogenic peptide substrate supplied with this kit is a proteasome substrate commonly used for measuring chymotrypsin-like peptidase activity. The detection of proteolytic activity is based on the release of free AMC fluorophore and offers the following advantages: non-radioactive, convenient real-time kinetics of substrate cleavage, and excellent sensitivity. A proteasome inhibitor, epoxomicin, is included as a test inhibitor for screening purposes. Applications: Fluorescent detection, HTS Activity assay.
Catalog No. 50-201-0300 Supplier Enzo Life Sciences Supplier No. BMLAK7400001
A QUANTIZYME Assay System. Fluorogenic, non-radioactive assay designed to measure chymotrypsin-like protease activity of purified 20S proteasome. This 96-well assay is useful for screening inhibitors and modulators of the 20S proteasome. The kit includes purified, human erythrocyte 20S proteasome (20µg). The Suc-LLVY-AMC fluorogenic peptide substrate supplied with this kit is a proteasome substrate commonly used for measuring chymotrypsin-like peptidase activity. The detection of proteolytic activity is based on the release of free AMC fluorophore and offers the following advantages: non-radioactive, convenient real-time kinetics of substrate cleavage, and excellent sensitivity. A proteasome inhibitor, epoxomicin, is included as a test inhibitor for screening purposes. Applications: Fluorescent detection, HTS Activity assay.