Learn More
Enzo Life Sciences Proteasome 20S beta2i subunit polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81500100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 218-236 of mouse proteasome 20S (β2i subunit). Species reactivity: Human, Mouse. Specificity: Recognizes the β2i subunit of the 20S proteasome. Applications: IF, IHC, WB. Formulation: Liquid. Antiserum containing 10mM sodium azide.
Catalog No. 50-201-1586