Learn More
Medchemexpress LLC Proteasome 20S C2/HC2 Antibody (YA2301) | 100 UL

Supplier: Medchemexpress LLC HYP82556100UL
Proteasome 20S C2/HC2 Antibody (YA2301) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting Proteasome 20S C2/HC2. It is for research use only.
- Rabbit-derived, non-conjugated IgG monoclonal antibody
- Targets proteasome 20S C2/HC2
- For research use only
- Predicted and observed band size: 30 kDa
- Purity by affinity chromatography
- Reactivity: Human, Mouse, Rat
- Host: Rabbit
- Isotype: IgG
- Applications: WB, IHC-P, ICC/IF, IP, FC
- Storage at -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.