Learn More
Enzo Life Sciences Proteasome 20S core subunits polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW93550100
Encompass_Preferred
Immunogen: Saccharomyces cerevisiae derived proteasomes. Species reactivity: Yeast. Specificity: Recognizes the 'core' subunits of the 20S proteasome. Applications: WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1681