Learn More
Enzo Life Sciences Proteasome 20S core subunits polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81550025
Encompass_Preferred
Host: Rabbit. Immunogen: Human erythrocyte-derived proteasomes. Source: From rabbit serum. Species Reactivity: Human, Mouse, Rat, Yeast, Rabbit. Specificity: Recognizes the 'core' subunits of the 20S proteasome. Applications: IHC (FS), IP, WB. Formulation: Liquid. In PBS containing 10mM sodium azide. Handling: Avoid freeze/thaw cycles. Aliquot undiluted antibody into smaller volumes (not less than 10µl) prior to freezing if appropriate. The use of high quality ‘antiserum-grade’ plastic or glass vials is recommended. Dilute to working strength with phosphate buffered saline pH 7.2-7.4 and 1% normal goat serum (if a goat anti-rabbit IgG linker antibody is to be used). Store diluted antibody at +2-4°C (do not freeze) and use within 1 month.
Catalog No. NC0603631