Learn More
Enzo Life Sciences Proteasome activator 11S alpha subunit polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81850025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 5-19 of mouse PA28α. Species reactivity: Human, Mouse, Rat. Specificity: Recognizes the α subunit of proteasome activator 11S. Applications: WB. Formulation: Liquid. Antiserum containing 10mM sodium azide.
Catalog No. 50-201-1589