Learn More
Apexbio Technology LLC PROTEASOME ACT FLU KIT-96mg
SDP
Catalog No. 5000567477
Encompass_Preferred
Proteasome Activity Fluorometric Assay Kit is a biochemical tool used in the field of cellular and molecular biology particularly in the study of protein degradation pathways Originating from research on the ubiquitin-proteasome system this assay is designed to measure proteasome activity which is crucial for regulating protein turnover within cells thereby maintaining cellular homeostasis The assay operates by detecting the release of a fluorophore from a labeled peptide substrate indicating proteolytic activity It is commonly applied in in vitro settings exhibiting activity in the nanomolar to low micromolar range Typical applications include studies using cell models to investigate proteasome inhibitors cancer research neurodegenerative disease models and drug screening programs The kit is suitable for exploring the effects of various compounds on proteasome function and experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67477 Supplier Apexbio Technology LLC Supplier No. K209696
Proteasome Activity Fluorometric Assay Kit is a biochemical tool used in the field of cellular and molecular biology particularly in the study of protein degradation pathways Originating from research on the ubiquitin-proteasome system this assay is designed to measure proteasome activity which is crucial for regulating protein turnover within cells thereby maintaining cellular homeostasis The assay operates by detecting the release of a fluorophore from a labeled peptide substrate indicating proteolytic activity It is commonly applied in in vitro settings exhibiting activity in the nanomolar to low micromolar range Typical applications include studies using cell models to investigate proteasome inhibitors cancer research neurodegenerative disease models and drug screening programs The kit is suitable for exploring the effects of various compounds on proteasome function and experimental concentrations typically depend on the specific research design