Learn More
Ubpbio Proteasome Activity Fluorometric Assay Kit II
Catalog No. 501965155
Encompass_Preferred
Proteasome Activity Fluorometric Assay Kit II can be used for assaying the constitutive proteasomes' chymotrypsin-like, trypsin-like and caspase-like activities in vitro using purified proteasome, cell lysates or tissue extracts. The proteasome cleaves the supplied fluorogenic substrates, and the released AMC fluorescence can be monitored using a plate reader or fluorometer at the excitation/emission wavelength of 360nm/460nm, respectively.
Catalog No. 50-196-5155 Supplier Ubpbio Supplier No. J4120
Proteasome Activity Fluorometric Assay Kit II can be used for assaying the constitutive proteasomes' chymotrypsin-like, trypsin-like and caspase-like activities in vitro using purified proteasome, cell lysates or tissue extracts. The proteasome cleaves the supplied fluorogenic substrates, and the released AMC fluorescence can be monitored using a plate reader or fluorometer at the excitation/emission wavelength of 360nm/460nm, respectively.