Learn More
Enzo Life Sciences Proteasome Assay Buffer (20ml)
SDP
Catalog No. 502011115
Encompass_Preferred
Assay buffer from the 20S Proteasome Assay Kit for Drug Discovery (BML-AK740). Formulation: Lyophilized. Contains 50mM Tris pH 7.5, 25mM KCl, 10mM NaCl, 1mM MgCl2. Long Term Storage: -20°C.
Catalog No. 50-201-1115 Supplier Enzo Life Sciences Supplier No. BMLKI3400020
Assay buffer from the 20S Proteasome Assay Kit for Drug Discovery (BML-AK740). Formulation: Lyophilized. Contains 50mM Tris pH 7.5, 25mM KCl, 10mM NaCl, 1mM MgCl2. Long Term Storage: -20°C.