Learn More
Enzo Life Sciences Proteasome inhibitor II (aldehyde) (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260090M005
Encompass_Preferred
Potent and cell permeable proteasome inhibitor. Inhibits the chymotrypsin-like activity of the 26S proteasome (MCP; multicatalytic proteinase complex). Does not inhibit the peptidylglutamyl-peptide hydrolyzing activity of the MPC. Alternative name: Z-LLF-CHO, Z-Leu-Leu-Phe-CHO. Formula: C29H39N3O5. MW: 509.7. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8312