Learn More
Medchemexpress LLC Proteasome inhibitor IX (PS-IX; AM114) | 856849-35-9 | 99.4% | 377.01 | C20H21B2NO5 | 10 MG
SDP
Catalog No. 5000239780
Encompass_Preferred
Proteasome inhibitor IX (PS-IX; AM114) is a chalcone-derived small molecule research reagent that inhibits the chymotrypsin-like activity of the 20S proteasome with an IC50 of approximately 1 μM. It is supplied as a light yellow solid intended for in vitro studies of proteasome function and anticancer mechanisms.
  • Inhibits chymotrypsin-like activity of 20S proteasome (IC50 ≈ 1 μM).
  • Chalcone-derived small molecule suitable for biochemical and cell-based assays.
  • High purity (99.4%) for reproducible results.
  • Molecular weight 377.01 and formula C20H21B2NO5.
  • Supplied as a light yellow solid for convenient handling and storage.
Catalog No. 50-002-39780 Supplier Medchemexpress LLC Supplier No. HY10741210MG
Proteasome inhibitor IX (PS-IX; AM114) is a chalcone-derived small molecule research reagent that inhibits the chymotrypsin-like activity of the 20S proteasome with an IC50 of approximately 1 μM. It is supplied as a light yellow solid intended for in vitro studies of proteasome function and anticancer mechanisms.
  • Inhibits chymotrypsin-like activity of 20S proteasome (IC50 ≈ 1 μM).
  • Chalcone-derived small molecule suitable for biochemical and cell-based assays.
  • High purity (99.4%) for reproducible results.
  • Molecular weight 377.01 and formula C20H21B2NO5.
  • Supplied as a light yellow solid for convenient handling and storage.