Learn More
Essendant PROTECSORCVRL2XLHDWH25
Catalog No. 5000022782
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees When your workers are in nonstop contact with liquid splashes and particles KleenGuard A40 Liquid and Particle Protection Coveralls are a great addition to offer your team comfortable protective clothing Features to provide more protection include the seamless front (for primary exposure area) and 1 inch zipper flap They feature REFLEX design which offers excellent fit and comfort (with elastic in the back) and freedom of movement Made of breathable microporous film to keep out debris non-hazardous liquid chemicals and dry particulates the KleenGuard A40 material gives your employees a better liquid barrier and a better particulate barrier than the leading competitor These coveralls with hood pass NFPA 99 for antistatic materials and pass ASTM F1670 testing for penetration of blood and bodily fluids Popular industries using this
Catalog No. 50-000-22782 Supplier Essendant Supplier No. ANS44325
Add to Cart
Add to Cart
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees When your workers are in nonstop contact with liquid splashes and particles KleenGuard A40 Liquid and Particle Protection Coveralls are a great addition to offer your team comfortable protective clothing Features to provide more protection include the seamless front (for primary exposure area) and 1 inch zipper flap They feature REFLEX design which offers excellent fit and comfort (with elastic in the back) and freedom of movement Made of breathable microporous film to keep out debris non-hazardous liquid chemicals and dry particulates the KleenGuard A40 material gives your employees a better liquid barrier and a better particulate barrier than the leading competitor These coveralls with hood pass NFPA 99 for antistatic materials and pass ASTM F1670 testing for penetration of blood and bodily fluids Popular industries using this