The PROTECTA® PRO™ Tongue Buckle Belt is designed with durable, corrosion-resistant hardware and features a quick tongue buckle adjustment for comfort during long workdays. It includes a back D-ring and two built-in grommeted miner's gear straps, providing hands-free convenience for construction, maintenance, and oil production tasks. This belt can be used independently or added to most harnesses for enhanced back support and versatility, ideal for restraint where no free fall is permissible.
- Back D-ring for connection to the restraint system
- Two built-in grommeted miner's gear straps
- Allows adding a hip pad or carrying tools and pouches
- Fast and easy tongue buckle adjustment
- Durable polyester construction
- High strength corrosion-resistant hardware
- Compact and lightweight design