Learn More
MSC PROTECTA Construction Safety Harness, X-Large, Body Belt, 1 ea
Catalog No. 5000796142
Encompass
This safety harness features comfort padding and durable polyester web construction. Designed for industrial workers, it provides optimal comfort and support for extended wear in harsh environments, ensuring reliable fall protection.
  • General purpose application
  • Body belt included
  • Pass-through chest connection
  • X-Large size
  • Back D-ring placement
  • Fixed dorsal D-ring to minimize readjustment
  • Integrated auto-setting lanyard keepers
  • Durable polyester web construction
  • Zinc plated steel buckles and D-rings
  • Weight capacity: 420 lb (190 kg)
Catalog No. 50-007-96142 Supplier MSC Supplier No. 23137425
This safety harness features comfort padding and durable polyester web construction. Designed for industrial workers, it provides optimal comfort and support for extended wear in harsh environments, ensuring reliable fall protection.
  • General purpose application
  • Body belt included
  • Pass-through chest connection
  • X-Large size
  • Back D-ring placement
  • Fixed dorsal D-ring to minimize readjustment
  • Integrated auto-setting lanyard keepers
  • Durable polyester web construction
  • Zinc plated steel buckles and D-rings
  • Weight capacity: 420 lb (190 kg)