Learn More
MSC Bell Laboratories Rodent Bait Station, Low Profile, Triangular, Tamper-Resistant
Catalog No. 502837300
Encompass
The Protecta LP is a triangular, rat-sized rodent station designed for corners and flush wall placement where rodents travel. Its low profile makes it suitable for indoor use under pallets and in tight baiting locations. Patented interior baffles guide rodents directly to the feeding receptacle. It is tamper-resistant, locking automatically when closed and unlocking with a special 2-prong key.
  • Patented low profile design fits corners and flush along walls.
  • Ideal for indoor use under pallets and in tight baiting locations.
  • Interior baffles guide rodents to the feeding receptacle.
  • Holds Blox, Super-Size Blox, tracking powders, liquid, cereal baits, and place pacs.
  • Horizontal and vertical bait rods secure bait.
  • Tamper-resistant with automatic locking.
  • Unlocks with a 2-prong key (compatible with other rodent stations).
  • Available in black and light gray.
Catalog No. 50-283-7300 Supplier MSC Supplier No. 74317389
The Protecta LP is a triangular, rat-sized rodent station designed for corners and flush wall placement where rodents travel. Its low profile makes it suitable for indoor use under pallets and in tight baiting locations. Patented interior baffles guide rodents directly to the feeding receptacle. It is tamper-resistant, locking automatically when closed and unlocking with a special 2-prong key.
  • Patented low profile design fits corners and flush along walls.
  • Ideal for indoor use under pallets and in tight baiting locations.
  • Interior baffles guide rodents to the feeding receptacle.
  • Holds Blox, Super-Size Blox, tracking powders, liquid, cereal baits, and place pacs.
  • Horizontal and vertical bait rods secure bait.
  • Tamper-resistant with automatic locking.
  • Unlocks with a 2-prong key (compatible with other rodent stations).
  • Available in black and light gray.