Learn More
3M™ PROTECTA™ PRO™ Confined Space System
Catalog No. 19809709 Shop All 3M Products
Change view
Click to view available options
Confined space system
Catalog No.
19-809-709 Confined space system
1 options
Catalog No. 19-809-709 Supplier 3M™ Supplier No. 70007492708
3M™ PROTECTA™ PRO™ Confined Space System features an 8 ft. (2.4 m) aluminum tripod with 50 ft. (15 m) galvanized 3-way self retracting lifeline, mounting bracket and pulley.
  • Complete confined space system
  • 8 ft. (2.4 m) PRO™ aluminum tripod
  • Telescoping adjustable locking legs
  • Protecta­™ 3-way self retracting lifeline with retrieval winch
  • 50 ft. (15.2 m) 3/16 in. (5 mm) galvanized steel cable lifeline
  • Pulley for confined space entry
  • Routes winch lifeline
  • Mounting bracket attaches SRL to tripod
  • Durable and compact design
TRUSTED_SUSTAINABILITY
Specifications
Weight (English) 74.2 kg
Weight (Metric) 163.58 kg
Certifications/Compliance OSHA 1926.502, OSHA 1910.146, OSHA 1910.66, ANSI Z117.1-1995
Product Type Confined Space System