Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ PROTECTA™ PRO™ Stretch 100% Tie-Off Shock Absorbing Lanyard

Catalog No. 19070709 Shop All 3M Products
Change view
Click to view available options
Material:
Zinc-plated Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-070-709 Zinc-plated Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-709 Supplier 3M™ Supplier No. 7100168875
  • $249.00
    $228.65 / Each of 1

    Save $20.35

    8% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Offers complete freedom of movement while reducing trip fall hazards, dragging, or snagging by expanding when taut and automatically contracting in reaction to movements. The inner core begins to extend and absorb energy once subjected to a fall and limits the fall arrest forces to safe levels.

  • 6 ft. (1.8 m) double-leg 100% tie-off single-leg lanyard
  • Stretches in reaction to your movements
  • Abrasion resistant polyester tubular webbing
  • User friendly self locking snap hook at center
  • Self-locking steel rebar hook at leg ends
  • Energy absorbing design limits fall arrest forces
  • Built-in impact indicator
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Zinc-plated Steel
Load Bearing Capacity (Metric) 59 to 141 kg
Webbing Material Polyester Tubular Web
Load Bearing Capacity (English) 130 to 310 lb.
Includes 6 ft. (1.8 m) double-leg 100% tie-off with elastic web and snap hook at center, steel rebar hooks at leg ends
Leg Configuration Double-Leg 100% Tie-Off
Connector Types Rebar Hook and Snap Hook
Product Type Variable/ Elastic Lanyard Type
Certifications/Compliance ANSI A10.32, OSHA 1926.502, OSHA 1910.66
Length (English) 6 ft.
Length (Metric) 1.8 m
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.