Learn More
MSC Protecta Retrieval Harness, Medium/Large, 420 lb, 1 ea
Catalog No. 5000796379
Encompass
This Protecta Vest-Style Retrieval Harness is designed for industrial and occupational use, featuring a fixed D-ring for consistent positioning and a modern, sleek design. It incorporates Quick-Connect Leg and Chest Connections and Back and Shoulder D-rings. The harness is rated for a total working weight of 420 lb (190 kg) and complies with ANSI Z359.11 and OSHA 1926.502 standards.
  • Low-profile suspension trauma straps integrated within webbing
  • Fixed D-ring minimizes readjustment during use
  • Auto-resetting lanyard keepers for fast parking of snap hooks and carabiners
  • Impact indicators allow easy inspection for prior damaging loads
  • Protected labels ensure long-term identification and inspection information
  • Quick-Connect Leg and Chest Connections
  • Back and Shoulder D-rings
Catalog No. 50-007-96379 Supplier MSC Supplier No. 16787988
This Protecta Vest-Style Retrieval Harness is designed for industrial and occupational use, featuring a fixed D-ring for consistent positioning and a modern, sleek design. It incorporates Quick-Connect Leg and Chest Connections and Back and Shoulder D-rings. The harness is rated for a total working weight of 420 lb (190 kg) and complies with ANSI Z359.11 and OSHA 1926.502 standards.
  • Low-profile suspension trauma straps integrated within webbing
  • Fixed D-ring minimizes readjustment during use
  • Auto-resetting lanyard keepers for fast parking of snap hooks and carabiners
  • Impact indicators allow easy inspection for prior damaging loads
  • Protected labels ensure long-term identification and inspection information
  • Quick-Connect Leg and Chest Connections
  • Back and Shoulder D-rings