Learn More
Bachem Fmoc-Dopa(acetonide)-OH
SDP
Supplier:  Bachem 4076682.0005
Encompass_Preferred
Fmoc-Dopa(acetonide)-OH, 852288-18-7, C27H25NO6, Mr 459.50, 5g. Fmoc-Dopa(isopropylidene ketal)-OH. Protected derivative for the introduction of Dopa during Fmoc-SPPS. Dopa-containing peptides adhere to metal surfaces. The catechol moiety forms complexes with metal ions, e.g. with Fe (III).
Catalog No. 50-194-7362