Learn More
Bachem Fmoc-4-(7-hydroxycoumarin-4-yl)-Abu-OH
SDP
Catalog No. 501947207
Encompass_Preferred
Fmoc-4-(7-hydroxycoumarin-4-yl)-Abu-OH, 1187744-84-8, C28H23NO7, Mr 485.49, 1g. Fmoc-(7-hydroxycoumarin-4-yl)-ethyl-Gly-OH, Fmoc-(umbellifer-4-yl)-ethyl-Gly-OH. Protected derivative of the fluorescent amino acid Cgl. Homolog of the coumarinylalanine derivative Fmoc-b-(7-methoxycoumarin-4-yl)-Ala-OH.
Catalog No. 50-194-7207 Supplier Bachem Supplier No. 4076431.1000
Fmoc-4-(7-hydroxycoumarin-4-yl)-Abu-OH, 1187744-84-8, C28H23NO7, Mr 485.49, 1g. Fmoc-(7-hydroxycoumarin-4-yl)-ethyl-Gly-OH, Fmoc-(umbellifer-4-yl)-ethyl-Gly-OH. Protected derivative of the fluorescent amino acid Cgl. Homolog of the coumarinylalanine derivative Fmoc-b-(7-methoxycoumarin-4-yl)-Ala-OH.