Learn More
Fritsch Milling And Sizing Inc Protected Funnel with Sample Pusher, P-19
SDP
Catalog No. 501534050
Encompass
The protected funnel with sample pusher for PULVERISETTE 19 enables due to the sample pusher a good handling and an outstanding material feeding.
Alternatively a standard funnel for long- and bulk solids is offered. This funnel is suitable to feed straw, film and other materials in their full length.
Catalog No. 50-153-4050 Supplier Fritsch Milling And Sizing Inc Supplier No. 19.1850.00
The protected funnel with sample pusher for PULVERISETTE 19 enables due to the sample pusher a good handling and an outstanding material feeding.
Alternatively a standard funnel for long- and bulk solids is offered. This funnel is suitable to feed straw, film and other materials in their full length.