Learn More
Fritsch Milling And Sizing Inc Protected Funnel with Sample Pusher, 3 Litres, P-25
SDP
Catalog No. 501534070
Encompass
The protected funnel with sample pusher for PULVERISETTE 25 enables due to the sample pusher a good handling and an outstanding material feeding.
Alternatively a standard funnel for long- and bulk solids is offered. This funnel is suitable to feed straw, film and other materials in their full length.
Catalog No. 50-153-4070 Supplier Fritsch Milling And Sizing Inc Supplier No. 25.2850.00
Add to Cart
Add to Cart
Request a Quote
The protected funnel with sample pusher for PULVERISETTE 25 enables due to the sample pusher a good handling and an outstanding material feeding.
Alternatively a standard funnel for long- and bulk solids is offered. This funnel is suitable to feed straw, film and other materials in their full length.