Learn More
Bachem Ac-a-benzyl-4,6-O-benzylidene-muramic acid
SDP
Supplier:  Bachem 4006354.1000
Encompass_Preferred
Ac-a-benzyl-4,6-O-benzylidene-muramic acid, 2862-03-5, C25H29NO8, Mr 471.51, 1g. Protected MurNAc derivative for the synthesis of acetyl-muramic acid peptides.
Catalog No. 50-194-7403