Learn More
Cayman Chemical ProtectIn D1 methyl ester 25ug
SDP
Catalog No. 502913873
Encompass_Preferred
An esterified form of protectin D1; an intermediate in the synthesis of protectin D1; reduces infarct area and volume and increases blood vessel density in the tissue surrounding the infarct in a rat model of MCAO-induced ischemic stroke
Catalog No. 50-291-3873 Supplier Cayman Chemical Supplier No. 3674625 UG
An esterified form of protectin D1; an intermediate in the synthesis of protectin D1; reduces infarct area and volume and increases blood vessel density in the tissue surrounding the infarct in a rat model of MCAO-induced ischemic stroke