Learn More
Cayman Chemical Protectin D1 Specialty Organic
SDP
Supplier:  Cayman Chemical 1001039025 UG
Encompass_Preferred
A specialized pro-resolving mediator; increases macrophage phagocytosis of PMNs in vitro and after 24 hours in vivo in mice; decreases PMN infiltration in a zymosan-induced mouse model of inflammation at 300 ng per animal; inhibits increases in neutrophil counts in BALF and lung myeloperoxidase activity in an LPS-induced mouse model of pulmonary injury and inflammation
Catalog No. 50-288-7607