Learn More
Cayman Chemical Protectin D1 Specialty Organic
SDP
Catalog No. 502887606
Encompass_Preferred
A specialized pro-resolving mediator; increases macrophage phagocytosis of PMNs in vitro and after 24 hours in vivo in mice; decreases PMN infiltration in a zymosan-induced mouse model of inflammation at 300 ng per animal; inhibits increases in neutrophil counts in BALF and lung myeloperoxidase activity in an LPS-induced mouse model of pulmonary injury and inflammation
Catalog No. 50-288-7606 Supplier Cayman Chemical Supplier No. 1001039010 UG
A specialized pro-resolving mediator; increases macrophage phagocytosis of PMNs in vitro and after 24 hours in vivo in mice; decreases PMN infiltration in a zymosan-induced mouse model of inflammation at 300 ng per animal; inhibits increases in neutrophil counts in BALF and lung myeloperoxidase activity in an LPS-induced mouse model of pulmonary injury and inflammation