Learn More
Anton Paar PROTECTIVE COVER FRONT (3 PCS) FOR 6.4" DISPLAY
Catalog No. NC3909508
Encompass_Preferred
Compatible with: Density and Sound Velocity Meter: DMA 6002 Sound Velocity, Density and Sound Velocity Meter: DMA 5001 Sound Velocity, Rolling-Ball Viscometer: Lovis 2000 M/ME, Analyzing system for alcoholic beverages: Alcolyzer ME, Soft Drink Analyzing System: Soft Drink Analyzer M, Density and Sound Velocity Meter: DSA 5000 M, Lovis 2001 Benchtop, Lovis 2001 Module
Catalog No. NC3909508 Supplier Anton Paar Supplier No. 88856
Compatible with: Density and Sound Velocity Meter: DMA 6002 Sound Velocity, Density and Sound Velocity Meter: DMA 5001 Sound Velocity, Rolling-Ball Viscometer: Lovis 2000 M/ME, Analyzing system for alcoholic beverages: Alcolyzer ME, Soft Drink Analyzing System: Soft Drink Analyzer M, Density and Sound Velocity Meter: DSA 5000 M, Lovis 2001 Benchtop, Lovis 2001 Module