Learn More
MSC Protective Eyewear, 3M, Black/Orange Frame, Gray Anti-Fog, 20 Ea
Catalog No. 5000788346
Encompass
The Privo Series protective eyewear offers comfortable and stylish eye protection. Designed for all-day wear, it features a soft nose bridge and temple tips for enhanced comfort. These unisex safety glasses have a sleek Euro-style appearance and are suitable for both indoor and outdoor use, with a sturdy polycarbonate lens for support.
  • Unisex safety glasses with a sleek Euro-style appearance and subtle lens wrap
  • Soft nose bridge and temple tips for all-day comfort
  • Polycarbonate lens absorbs 99.9% of UV rays
  • Anti-fog lens coating
  • Meets impact-rated protector requirement of ANSI-ISEA Z87.1-2020 standard
Catalog No. 50-007-88346 Supplier MSC Supplier No. 85684892
The Privo Series protective eyewear offers comfortable and stylish eye protection. Designed for all-day wear, it features a soft nose bridge and temple tips for enhanced comfort. These unisex safety glasses have a sleek Euro-style appearance and are suitable for both indoor and outdoor use, with a sturdy polycarbonate lens for support.
  • Unisex safety glasses with a sleek Euro-style appearance and subtle lens wrap
  • Soft nose bridge and temple tips for all-day comfort
  • Polycarbonate lens absorbs 99.9% of UV rays
  • Anti-fog lens coating
  • Meets impact-rated protector requirement of ANSI-ISEA Z87.1-2020 standard