Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Protective Eyewear Holder
SDP

Catalog No. 01259133
Change view
Click to view available options
Product Type:
Goggle Holder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
01-259-133 Goggle Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-133 Supplier Poltex Supplier No. PROTEYE12M
Add to Cart
Add to Cart

Holds 12 pairs of safety glasses, keeping them up and out of the way. Made from super-durable PETG with a magnet mount and powerful magnets included.

  • Optional mounting choices include wall mount with screws/anchors or HB tape mount with very high bond adhesive.
  • Supplied without eyewear, offering flexibility to use with any safety glasses brand or model.
  • Designed for long-lasting strength and reliable performance.

Specifications

Color Clear
Product Type Goggle Holder
For Use With (Equipment) Protective Eyeglasses
Dimensions (L x W x H) 12 x 6.5 x 8.5 in.
Length (English) 12 in.
Weight (English) 2 lb.
Material PETG
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.