Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Protective Eyewear Holder
SDP

Catalog No. 22000339
Change view
Click to view available options
Description:
Magnet Mount
Tape Mount
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
22-000-339 Tape Mount
22-000-338 Magnet Mount
22-000-337 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-339 Supplier Poltex Supplier No. MINIEYET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Holds and dispenses bulk protective eyewear in a compact design.

Durable PETG Construction.

Specifications

Height (English) 8.6 in.
Width (English) 6.3 in.
Description Tape Mount
Depth (English) 5.8 in.
Material PETG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.