Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Protective Eyewear Holder
SDP

Catalog No. 22000634
Change view
Click to view available options
Description:
50 Position
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
22-000-634 50 Position
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-634 Supplier Poltex Supplier No. PROTEYE50W
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Constructed from super-durable PETG, this Protective Eyewear Holder holds 50 pairs of safety glasses up and out of the way.

  • Keyhole slots for wall mount (screws/anchors included).

Specifications

Description 50 Position
Material PETG
Mounting Wall Mount
Positions 50 Positions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.