Learn More
MSC Protective Industrial Products Kevlar Latex Glove, Cut A4, 1 Pair
Catalog No. 5000684912
Encompass
This PowerGrab KEV seamless knit glove offers advanced protection against cut, puncture, and abrasion. Featuring a Kevlar liner and a latex coating with MicroFinish grip on the palm and fingers, it provides excellent dexterity and comfort for various industrial applications.
  • Kevlar fiber shell for superior cut resistance
  • Latex coated palm and fingers with MicroFinish grip
  • ANSI/ISEA Cut Level A4, Puncture Level 4, Abrasion Level 3
  • EN 388: 3X43D and EN 407 Contact Heat Level 2
  • Seamless knit construction, 10 gauge, with knit wrist cuff
  • Designed for durability and enhanced grip in dry and wet conditions
Catalog No. 50-006-84912 Supplier MSC Supplier No. 40380883
This PowerGrab KEV seamless knit glove offers advanced protection against cut, puncture, and abrasion. Featuring a Kevlar liner and a latex coating with MicroFinish grip on the palm and fingers, it provides excellent dexterity and comfort for various industrial applications.
  • Kevlar fiber shell for superior cut resistance
  • Latex coated palm and fingers with MicroFinish grip
  • ANSI/ISEA Cut Level A4, Puncture Level 4, Abrasion Level 3
  • EN 388: 3X43D and EN 407 Contact Heat Level 2
  • Seamless knit construction, 10 gauge, with knit wrist cuff
  • Designed for durability and enhanced grip in dry and wet conditions