Learn More
MSC Protective Industrial Products Neoprene Gloves, Raised Diamond Grip, 28 Mil
Catalog No. 5000766229
Encompass
Neoprene gloves provide superior protection from a wide range of chemicals such as acids, caustics, solvents and oils. They are ideal for use in the petrochemical industry, janitorial, general maintenance, and photo finishing.
  • Straight cuff for forearm protection from liquid run-off
  • Flock lining for increased comfort and ease of donning
  • Raised diamond pattern for enhanced grip
  • Made from component materials that comply with Federal regulations for food contact 21 CFR, 170-199
Catalog No. 50-007-66229 Supplier MSC Supplier No. 93415610
Neoprene gloves provide superior protection from a wide range of chemicals such as acids, caustics, solvents and oils. They are ideal for use in the petrochemical industry, janitorial, general maintenance, and photo finishing.
  • Straight cuff for forearm protection from liquid run-off
  • Flock lining for increased comfort and ease of donning
  • Raised diamond pattern for enhanced grip
  • Made from component materials that comply with Federal regulations for food contact 21 CFR, 170-199