CleanTeam single-use nitrile gloves are designed for use in controlled environments such as biotechnology, medical device manufacturing, and computer assembly. These ambidextrous, powder-free gloves offer superior tactile sensitivity and a secure grip for critical applications.
- Used in biotechnology, medical device manufacturing, and computer assembly.
- Exceeds standards required for use in Class 10 and other controlled environments.
- Finger textured for enhanced grip in wet or dry conditions.
- Ambidextrous design for quick and easy donning.
- Powder-free for clean comfort and superior tactile sensitivity.
- Beaded cuff enhances strength and prevents liquid runoff.
- Chlorinated surface for improved hardness and cleanliness.
- Electrostatic dissipative (ESD) properties for electronics assembly (109 Ohms).
- Silicone-free construction.