Learn More
Grainger Protective Leg Tip SlipOn Rd 1-1/8 PK25
Catalog No. 502765464
Encompass_Preferred
The 16ZF59 is a 16-gallon wet/dry vacuum made by Shop-Vac. It has a 6.5 peak horsepower motor and is designed for heavy-duty use. The vacuum has a large capacity stainless steel tank and is equipped with an onboard hose, a variety of attachments including a crevice tool, utility nozzle, extension wands and a cartridge filter. It can pick up both wet and dry debris. The vacuum has non-marking wheels and a rear blower port to make cleaning easy. It's made in the USA and comes with a 3 year warranty.
Catalog No. 50-276-5464 Supplier Grainger Supplier No. 16ZF59
The 16ZF59 is a 16-gallon wet/dry vacuum made by Shop-Vac. It has a 6.5 peak horsepower motor and is designed for heavy-duty use. The vacuum has a large capacity stainless steel tank and is equipped with an onboard hose, a variety of attachments including a crevice tool, utility nozzle, extension wands and a cartridge filter. It can pick up both wet and dry debris. The vacuum has non-marking wheels and a rear blower port to make cleaning easy. It's made in the USA and comes with a 3 year warranty.