Learn More
Grainger Protective Leg Tip SlipOn Rd 15/64 PK10
Catalog No. 502768022
Encompass_Preferred
The Grainger Approved Protective Leg Tip Slip-On is a safety product designed to help prevent injuries. It slips over the legs of ladders, scaffolding, and other equipment to cushion impacts and prevent punctures. The tip is made of durable rubber and has a diameter of 1-3/4". It features a ribbed interior to ensure a tight, secure fit over metal legs up to 1-1/2" in diameter. The slip-on design makes it easy to install without tools. This leg tip provides an economical way to help improve jobsite safety.
Catalog No. 50-276-8022 Supplier Grainger Supplier No. 16ZD84
The Grainger Approved Protective Leg Tip Slip-On is a safety product designed to help prevent injuries. It slips over the legs of ladders, scaffolding, and other equipment to cushion impacts and prevent punctures. The tip is made of durable rubber and has a diameter of 1-3/4". It features a ribbed interior to ensure a tight, secure fit over metal legs up to 1-1/2" in diameter. The slip-on design makes it easy to install without tools. This leg tip provides an economical way to help improve jobsite safety.