Learn More
Grainger Protective Leg Tip SlipOn Rd 19/32 PK25
Catalog No. 502765272
Encompass_Preferred
The Grainger 16ZE10 is a Westward brand 1/4-inch drive socket set. It contains 10 sockets ranging in size from 4mm to 14mm. The sockets are made from chrome vanadium steel and have a black phosphate finish. The set comes in a molded plastic case for storage and portability. This socket set is ideal for automotive and light industrial applications that require standard and deep metric sockets in the most common sizes. The lifetime warranty provides peace of mind. With durable construction and useful sizes, this socket set is a versatile addition to any toolbox.
Catalog No. 50-276-5272 Supplier Grainger Supplier No. 16ZE10
The Grainger 16ZE10 is a Westward brand 1/4-inch drive socket set. It contains 10 sockets ranging in size from 4mm to 14mm. The sockets are made from chrome vanadium steel and have a black phosphate finish. The set comes in a molded plastic case for storage and portability. This socket set is ideal for automotive and light industrial applications that require standard and deep metric sockets in the most common sizes. The lifetime warranty provides peace of mind. With durable construction and useful sizes, this socket set is a versatile addition to any toolbox.