Learn More
Grainger Protective Leg Tip SlipOn Rd 1-1/4 PK25
Catalog No. 502765317
Encompass_Preferred
The Grainger Approved CTB-22B Furniture Protective Leg Tips are designed to protect floors from scratches and damage caused by furniture legs. They are made of clear thermoplastic rubber and have a dome shape to provide full floor contact. The leg tips have an inner diameter of 0.375 inches and an outer diameter of 0.75 inches. They are sold in packs of 4 tips per pack. The leg tips are easy to install by pressing onto the bottom of furniture legs. Overall, the CTB-22B leg tips provide a low-cost way to protect hardwood, tile, and other flooring types.
Catalog No. 50-276-5317 Supplier Grainger Supplier No. 16ZF63
The Grainger Approved CTB-22B Furniture Protective Leg Tips are designed to protect floors from scratches and damage caused by furniture legs. They are made of clear thermoplastic rubber and have a dome shape to provide full floor contact. The leg tips have an inner diameter of 0.375 inches and an outer diameter of 0.75 inches. They are sold in packs of 4 tips per pack. The leg tips are easy to install by pressing onto the bottom of furniture legs. Overall, the CTB-22B leg tips provide a low-cost way to protect hardwood, tile, and other flooring types.