Learn More
Grainger Protective Leg Tip SlipOn Rd 23/64 PK10
Catalog No. 502766140
Encompass_Preferred
The 16ZD88 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-B:C and contains 1.4 gallons of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It features an all-steel valve construction with stainless steel valve stems. The cylinder is made of steel that is coated with red enamel paint. This extinguisher has a discharge range of 10-15 feet and discharges in 10 seconds. It is intended for commercial applications. The extinguisher weighs approximately 26 lbs when full and measures 20.5" tall x 7.75" diameter.
Catalog No. 50-276-6140 Supplier Grainger Supplier No. 16ZD88
The 16ZD88 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-B:C and contains 1.4 gallons of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It features an all-steel valve construction with stainless steel valve stems. The cylinder is made of steel that is coated with red enamel paint. This extinguisher has a discharge range of 10-15 feet and discharges in 10 seconds. It is intended for commercial applications. The extinguisher weighs approximately 26 lbs when full and measures 20.5" tall x 7.75" diameter.